Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Stainless-Steel Forceps
Nalgene Stainless-Steel Forceps are available in bent or straight-tip styles. Bent-tip style is designed for easy removal and manipulation of filter membranes.
$122.45 - $122.81
Specifications
No. per Case 1
No. per Pack 1
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Tip Style Price Quantity    
09-742
thermo scientific™
NDS03990001
Bent tip Each for $122.81
09-744
thermo scientific™
DS03990002
Straight tip Case for $122.45
Description & Specifications
Specifications
No. per Case 1
No. per Pack 1
  • Stainless steel
  • Self-closing (bent tip)
  • Serrated gripping area
  • Autoclavable
Warranty: 90 days