Thermo Scientific™ Nalgene™ Stainless-Steel Forceps, straight tip
Nalgene Stainless-Steel Forceps are available in bent or straight-tip styles. Bent-tip style is designed for easy removal and manipulation of filter membranes.
Supplier: Thermo Scientific™ DS03990002
Description
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
- Stainless steel
- Self-closing (bent tip)
- Serrated gripping area
- Autoclavable
Specifications
Straight, Flat, Serrated | |
Stainless Steel | |
Forceps | |
Ea. |
1 | |
Forceps | |
Stainless-Steel Forceps, straight tip | |
Straight |