Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Square Wide-Mouth Large PPCO Bottle with Closure: Autoclavable
Autoclave and store contaminated slides and other small lab equipment with the Thermo Scientific™ Nalgene™ Square Wide-Mouth Large PPCO Bottle with Closure.
$77.81 - $77.81
Products
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity    
03-313
thermo scientific™
21220010
4L 100mm Each for $77.81
Case of 6 Each for $466.86
Description & Specifications
Autoclave and store contaminated slides and other small lab equipment with the Thermo Scientific™ Nalgene™ Square Wide-Mouth Large PPCO Bottle with Closure. This polypropylene copolymer bottle is excellent for collecting, storing and transporting dry chemicals and environmental samples. Space saving design features molded-in handgrips for greater convenience and safety when pouring.
Featres:
  • Polypropylene screw closures included
  • Wide mouth makes it excellent for collecting, storing and transporting dry chemicals and environmental samples
  • Bottle is not liquid leakproof†
Note:
Completely disengage threads or remove cap before autoclaving.
<
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.