Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Square PET Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Provide excellent gas barrier properties for storage and shipping of liquid media, buffers and sera with these square bottles.
$256.51 - $287.55
Specifications
Certifications/Compliance PET Media Bottles and HDPE closure materials meet current biological testing standards including USP Class VI, USP <661>, European Pharmacopeia for Abnormal Toxicity
Closure Size 38-430mm
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28mm
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. �The term �leakproof� applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity    
02-925-00A
thermo scientific™
342040-0650
650mL 38-430mm Case of 48 Each for $287.55
02-925-00B
thermo scientific™
N3420401000*
1L 38-430mm Case of 24 Each for $256.51
Description & Specifications
Specifications
Certifications/Compliance PET Media Bottles and HDPE closure materials meet current biological testing standards including USP Class VI, USP <661>, European Pharmacopeia for Abnormal Toxicity
Closure Size 38-430mm
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28mm
Provide excellent gas barrier properties for storage and shipping of liquid media, buffers and sera with Thermo Scientific™ Nalgene™ Square PET Media Bottles with Closure. These light-weight bottles are shatter-resistant, sterile and come in a space-saving square design.
Features:
  • Polyethylene terephthalate with natural high density polyethylene closure
  • Molded in graduations and neck ring for use with tamper evident shrink sleeves
  • Gamma irradiated to 10-6 Sterility Assurance Level (SAL)
  • Non-pyrogenic and non-cytotoxic
  • Transparency and reduced permeability to CO2 and O2
  • Bottle and closure material are Animal Derived Component Free (ADCF)
  • Leakproof†
Ordering Information:
Products are packaged in shrink-wrap trays. Trays are double polybagged inside the master carton.