Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Spherical-Bottom Polycarbonate Centrifuge Bottle
Process large-volume cell harvesting, pelleting and protein purification with this centrifuge bottle. A spherical-bottom design facilitates pellet formation and retrieval.
$230.27 - $230.27
Products
Catalog Number Mfr. No. Max. Force Nominal Capacity Price Quantity    
05-579
thermo scientific™
31230250
27,500 x G 250mL Pack of 4 Each for $230.27
Case of 9 Pack for $2,072.43
Description & Specifications
Ideal for large-volume cell harvesting, pelleting and protein purification, Thermo Scientific™ Nalgene™ Spherical-Bottom Polycarbonate Centrifuge Bottles are the perfect fit with Thermo Scientific™ Fiberlite™ carbon fiber rotors. Molded of transparent polycarbonate, these translucent bottles are autoclavable and have excellent mechanical strength properties. The spherical-bottom design facilitates pellet formation and retrieval.
  • Molded of transparent polycarbonate for easy visualization of samples
  • Spherical-bottom design facilitates pellet formation and retrieval
  • For proper performance, bottles must be filled to 80% of the total capacity
  • For use in standard rotors, the use of the Nalgene spherical-bottom adaptor DS3124-0010 is recommended
  • For low to moderate speed centrifugation of biological and chemical samples
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.