Thermo Scientific™ Nalgene™ Settlometer Kit
Analyze the activated sludge process using this all-plastic tool for waste water treatment plant operators, a cost-effective alternative when compared to glass equivalents.
$318.45 - $318.45
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
15-443-35 Thermo Scientific™
N10100507
Settlometer Kit Each for $318.45
Case of 4 Each for $1,273.80
Specifications
Description
Analyze the activated sludge process using the Thermo Scientific™ Nalgene™ Settlometer Kit. An all-plastic tool for waste water treatment plant operators, this kit is a cost-effective alternative to glass equivalents. Settlometer and centrifuge tests for sludge quality are used in analyzing the activated sludge process. The 2L jar is lightweight, transparent, highly impact-resistant and has easy to read white graduations. The paddle permits agitation without scratching jar. Tubes fit tabletop centrifuges up to 6000 x g without cushions or adapters.
Features:
  • 2L jar is lightweight, transparent, and impact-resistant with easy-to-read white graduations
  • Paddle permits agitation of sample without scratching jar
  • Centrifuge tubes feature molded-in graduations and friction-fit closures
  • For tabletop (clinical) centrifuges; shatterproof tubes will withstand forces up to 6000 × g without cushions or adapters, eliminating risk of broken glass
Ordering Information:
  • Approximate jar dimensions: O.D. × H: 3.3 × 7.5″ (13.2 × 19.1cm)
  • Tube dimensions, O.D. × H: 0.68 × 25.6″ (1.7 × 11.9cm)
  • Replacement jar and lid available, order Cat. No. 711010-9507
Includes:
  • Polycarbonate jar and cover
  • Polypropylene paddle
  • Ten 0.5 oz. (15mL) polycarbonate conical centrifuge tubes with friction-fit polypropylene closures
  • Instruction booklet
  • Data sheet
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.