missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Round Polycarbonate Clearboy™ with Spigot

Prepare large volume media and cultures with these round polycarbonate Clearboys. Lighter, safer and more durable than glass. Useful for visual quality inspections.

$490.16 - $696.74

Specifications

Diameter (Metric) Inner Neck 66mm
Autoclavable Yes
Closure Size 83B
Graduations Molded-in graduations in 1 gal. and 5L increments
Material Polycarbonate
View More Specs

Disclaimers:

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
02-962-7G Thermo Scientific™
N23170020
10L 83B Each for $490.16
Case of 4 Each for $1,960.64
02-962-7H Thermo Scientific™
N23170050
20L 83B Each for $696.74
Case of 4 Each for $2,786.96
Specifications

Description

Prepare large volume media and cultures with Thermo Scientific™ Nalgene™ Round Polycarbonate Clearboy™ with Spigot. These durable clearboys are lighter and safer than glass, and especially useful for visual inspection of contents for quality. Ideal for refrigerated or frozen storage of aqueous solutions, or for dispensing sterile water, aqueous reagents and media.

Features:
  • Molded in graduations in 1 gal and 5L increments for ease in dispensing
  • Molded in spigot boss provides strength and leak resistance
  • Polycarbonate is glass-clear for easy visual inspection and volume measurements
  • Suitable for mixing, storing and freezing aqueous-based reagents and biologicals
  • Designed with 83mm neck to accept TopWorks and other Nalgene accessories for versatility
  • Graduated
  • Leakproof†
  • Autoclavable for sterile applications 

Includes:

Polypropylene spigot screw closure and TPE gasket. See autoclaving instructions for more information.

Note:

Completely disengage closure threads and disassemble spigot before autoclaving
Follow product insert instructions for autoclaving to ensure proper function
Note: Tubing adaptor is not autoclavable

Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com

Description & Specifications

Specifications

66mm
83B
Polycarbonate
4
Yes
Molded-in graduations in 1 gal. and 5L increments
Yes
1