Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Round Polycarbonate Clearboy™ with Closure
Conduct large-volume media and culture preparation with these polycarbonate Nalgene Clearboys, particularly when visual inspection is required.
$413.77 - $590.74
Specifications
Description Round Transparent Clearboy
Autoclavable Yes
Closure Size 83B
Handles No
Material Polycarbonate
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity    
02-962-6A
thermo scientific™
N22510020
10L 83B Each for $413.77
Case of 4 Each for $1,655.08
02-962-6B
thermo scientific™
N22510050
20L 83B Each for $590.74
Case of 4 Each for $2,362.96
Description & Specifications
Specifications
Description Round Transparent Clearboy
Autoclavable Yes
Closure Size 83B
Handles No
Material Polycarbonate
Spigot No
No. per Case 4
No. per Pack 1
Conduct large-volume media and culture preparation with these polycarbonate Nalgene Clearboys, particularly when visual inspection is required. This polycarbonate Clearboy is durable, lighter and safer than glass, autoclavable for sterile applications and ideal for refrigerated or frozen storage of aqueous solutions such as sterile water, aqueous reagents and media.
Features:
  • Available in 10 and 20L sizes for a variety of biopharmaceutical applications
  • Graduated in liters and gallons
  • Molded of transparent, durable polycarbonate
  • Customization options available for fluid transfer applications
  • Autoclavable/Graduated/Leakproof†
Includes:
  • Polypropylene closure and TPE gasket
Ordering Information:
Additional closure options include Nalgene Quick Filling/Venting Closures (Cat. No. 2158), Nalgene Filling/Venting Closures(Cat. No. 2162) Nalgene Top Works™ Fluid Transfer Systems (Cat. No. 2135) and Nalgene Validation Bottles (Cat. No. DS2127). Additional accessories include Nalgene Barbed Bulk Head Fittings (Cat. No. 6149).
Warranty: 90 days
Note:
Completely disengage closure threads and disassemble spigot before autoclaving.
The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. B) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.