Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Round LDPE Carboys
Store and transport reagents with the Thermo Scientific™ Nalgene™ Round LDPE Carboys, featuring wide shoulder handles for easy carrying and pouring.
$181.32 - $553.16
Specifications
Diameter (Metric) Inner Neck 64mm
Closure Size 83B
Handles Yes
Material LDPE
No. per Pack 1
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products
Catalog Number Mfr. No. Description Capacity (Metric) Closure Size Price Quantity    
02-961D
thermo scientific™
N22100020
Round LDPE Carboy w/handle 10L 83B Each for $181.32
Case of 6 Each for $1,087.92
02-961E
thermo scientific™
N22100040
Round LDPE Carboy w/handle 15L 83B Each for $225.77
Case of 4 Each for $903.08
02-961A
thermo scientific™
N22100050
Round LDPE Carboy w/handle 20L 83B Each for $258.84
Case of 4 Each for $1,035.36
02-961B
thermo scientific™
N22100065
Round LDPE Carboy w/handle 25L 83B Each for $319.26
Case of 4 Each for $1,277.04
02-961C
thermo scientific™
N22100130
Round LDPE Carboy w/handle 50L 83B Each for $553.16
Description & Specifications
Specifications
Diameter (Metric) Inner Neck 64mm
Closure Size 83B
Handles Yes
Material LDPE
No. per Pack 1
Store and transport reagents with the Thermo Scientific™ Nalgene™ Round LDPE Carboys, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands. These carboys accept 13.5 rubber stoppers for an alternative sealing method.
  • Molded-in graduations of 5L and 1 gal. increments
  • Leakproof†
Includes:
White polypropylene screw cap and 83B closure
Ordering Information:
8210 Series is manufactured in our state-of-the-art Suzhou, China facility and is available for purchase in Asia Pacific region.  All other products manufactured in Rochester, NY.