Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Round LDPE Carboys with Spigot
Collect and dispense distilled water, acids and reagents with Thermo Scientific™ Nalgene™ Round LDPE Carboys with Spigot.
$235.03 - $659.66
Specifications
Description Round Carboys w/Spigot
Graduations Molded-in graduations in 1 gal. and 5L increments
Handles Integral shoulder handles
Material LDPE
Spigot Molded-in spigot boss
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity    
02-963AA
thermo scientific™
N23180010
4L 38-430mm Each for $235.03
Case of 6 Each for $1,410.18
02-963A
thermo scientific™
N23180020
10L 83B Each for $317.84
Case of 6 Each for $1,907.04
02-963BB
thermo scientific™
N23180050
20L 83B Each for $373.16
Case of 4 Each for $1,492.64
02-963B
thermo scientific™
N23180065
25L 83B Each for $436.59
02-963C
thermo scientific™
N23180130
50L 83B Each for $659.66
Description & Specifications
Specifications
Description Round Carboys w/Spigot
Graduations Molded-in graduations in 1 gal. and 5L increments
Handles Integral shoulder handles
Material LDPE
Spigot Molded-in spigot boss
Collect and dispense distilled water, acids and reagents with Thermo Scientific™ Nalgene™ Round LDPE Carboys with Spigot. These low density polyethylene carboys have a spigot boss molded in for convenience, strength and leak resistance.
Features:
  • Molded in graduations of 1 gal. and 5L increments for ease in dispensing
  • Integral shoulder handles that make lifting comfortable for four largest sizes
  • Molded-in spigot boss for strength and leak resistance
  • Leakproof†
Ordering Information:
Supplied with shoulder loop on 1 gal. (3.8L) size, shoulder handles on all other sizes. 1 gal. size is not graduated.
Includes:
Polypropylene spigot and screw closure
Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com.