Learn More
Thermo Scientific™ Nalgene™ LDPE, Round Carboy with Spigot
For Use With: Collect and dispense distilled water, acids and reagents; Closure Material: Polypropylene; Color: White
Supplier: Thermo Scientific™ 23180130
Description
Thermo Scientific™ Nalgene Round LDPE Carboys are the icon of lab water dispensing and storage. Place one at every sink and every work station to enhance your productivity. Carboys have a spigot boss molded in once piece with the carboy for convenience, strength and leak resistance. Suitable for collecting and dispensing distilled water, acids and reagents. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.
- Molded in graduations in 1 gal. and 5L increments for ease in dispensing
- Integral shoulder handles make lifting comfortable for the four largest sizes
- Molded-in spigot boss provides strength and leak resistance
- Leakproof†
Ordering Information:
Supplied with shoulder loop on 1 gal. (3.8L) size, shoulder handles on all other sizes. 1 gal. size is not graduated.
Includes:
Polypropylene spigot and screw closure with TPE gasket † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes; b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Carboy with Spigot | |
Polypropylene | |
Round Carboys w/Spigot | |
379 mm | |
LDPE | |
Molded-in graduations in 1 gal. and 5 L increments | |
Integral shoulder handles, Molded-in spigot boss | |
White (Opaque) |
13 gal. | |
83 B | |
64 mm | |
50 L | |
Collect and dispense distilled water, acids and reagents | |
678 mm | |
Not Autoclavable |