Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Rectangular Polypropylene Carboys with Closure
Store media, solutions and sterile water in these rectangular polypropylene carboys, featuring a space-saving design and wide mouth opening for filling and cleaning.
$292.61 - $418.13
Specifications
Closure Size 100-415mm
Description Autoclavable Rectangular Carboys
Autoclavable Yes
Material Polypropylene
No. per Pack 1
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes; b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products
Catalog Number Mfr. No. Closure Size Capacity (Metric) Price Quantity    
02-961-53A
thermo scientific™
N22120020
100-415mm 9L Each for $292.61
Case of 6 Each for $1,755.66
02-961-53C
thermo scientific™
N22120050
100-415mm 20L Each for $418.13
Case of 4 Each for $1,672.52
Description & Specifications
Specifications
Closure Size 100-415mm
Description Autoclavable Rectangular Carboys
Autoclavable Yes
Material Polypropylene
No. per Pack 1
Store solutions and handle large volumes of powders and other solid samples with Thermo Scientific™ Nalgene™ Rectangular Polypropylene Carboys with Closure. Featuring a space-saving design, a wide mouth opening for filling and cleaning and a sturdy stainless-steel handle, these carboys are tough, translucent and autoclavable.
Features:
  • Wide-mouth opening for easy filling, transferring and cleaning of carboys
  • Molded in graduations in 1 gallon and 5L increments for ease in dispensing
  • Stainless-steel handle for ease in transporting containers
  • Autoclavable, graduated and leakproof†
Includes:
  • Polypropylene screw closure
Warranty: 90 days