Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ with Closure
Store media, solutions and sterile water in Nalgene Rectangular Clearboys. Featuring a space-saving design and extreme impact resistance. Ideal for cold room use.
$545.42 - $747.06
Specifications
Closure Size 100-415mm
Description Rectangular Clearboy
Autoclavable Yes
Material Polycarbonate
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products
Catalog Number Mfr. No. Closure Size Capacity (Metric) Price Quantity    
02-961-55A
thermo scientific™
NDS22130020
100-415mm 9L Each for $545.42
02-961-55C
thermo scientific™
NDS22130050
100-415mm 20L Each for $747.06
Description & Specifications
Specifications
Closure Size 100-415mm
Description Rectangular Clearboy
Autoclavable Yes
Material Polycarbonate
Store media, solutions and sterile water in Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboys with Closure. Featuring a space-saving design, a wide mouth opening for filling and cleaning and a sturdy stainless-steel handle. The clearboy offers extreme impact resistance, and is ideal for cold room use.
Features:
  • Molded in graduations in 1 gallon and 5L increments
  • Autoclavable/Transparent/Graduated/Leakproof†
Includes:
Polypropylene screw closure
Warranty: 90 days.