Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Rectangular HDPE Carboys with Tubulation and Closure
Save benchtop space with these HDPE carboys, molded in one piece with serrated tubulation. They are strong, leak-resistant and durable and accept 1/2″ ID tubing.
$314.68 - $314.68
Products
Catalog Number Mfr. No. Closure Size Capacity (Metric) Price Quantity    
02-962-5B
thermo scientific™
N23030050
100mm 20L Each for $314.68
Case of 4 Each for $1,258.72
Description & Specifications
Save benchtop space with Thermo Scientific™ Nalgene™ Rectangular HDPE Carboys with Tubulation and Closure, which are molded in one piece with serrated tubulation. They are strong, leak-resistant and durable and accept 1/2″ (13mm) ID tubing.
Features:
  • Wide-mouth opening for easy filling, transferring and cleaning of carboys
  • Molded-in graduations in 1 gal. and 5L increments for ease in dispensing
  • Molded in one piece for strength, leak resistance and a longer life
  • Low outlet that provides for more complete drainage
  • Molded-in handle on larger sizes for ease in transporting containers
  • Graduated
Includes:
  • Polypropylene screw cap
Note:
Hose clamp should be used when attaching tubing to ensure a tight fit and avoid leaking. Containers can be used for long-term storage, but direct UV exposure should be avoided.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.