Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Rectangular HDPE Carboy with Spigot

Store and dispense media, solutions and sterile water with space-saving Thermo Scientific™ Nalgene™ Rectangular HDPE Carboys.

$313.97 - $418.45

Specifications

Closure Size 100mm
Description Rectangular Carboy w/Spigot
Diameter (Metric) Inner Neck 86mm
Material HDPE
Spigot Molded-in spigot boss
View More Specs

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Products
Catalog Number Mfr. No. Closure Size Capacity (Metric) Price Quantity    

02-963-6A

 
thermo scientific™
N23200020
100mm 9L Each for $313.97
Case of 6 Each for $1,883.82

02-963-6B

 
thermo scientific™
N23200050
100mm 20L Each for $418.45
Case of 4 Each for $1,673.80
Description & Specifications

Specifications

Closure Size 100mm
Description Rectangular Carboy w/Spigot
Diameter (Metric) Inner Neck 86mm
Material HDPE
Spigot Molded-in spigot boss
No. per Pack 1

Store and dispense media, solutions and sterile water with space-saving Thermo Scientific™ Nalgene™ Rectangular HDPE Carboys with Spigot. These carboys feature a stainless-steel handle and a wide-mouth opening for filling and cleaning. Tough and rigid with excellent chemical resistance, this carboy is a good choice for dispensing stock solutions.

Features:

  • Polypropylene spigot and screw closure
  • Wide-mouth opening permits easy filling, transferring and cleaning of carboys
  • Molded-in graduations in 1 gallon and 5L increments for ease in dispensing
  • Stainless-steel handle for ease in transporting containers
  • Molded-in spigot boss provides strength and leak resistance
  • Spigot comes with a convenient tubing adapter providing versatility
  • Rectangular shape saves valuable space on your benchtop
  • 20L size has molded-in hand grips in the sidewall for easier handling
  • Graduated

Includes:

  • Polypropylene screw cap
  • Steel handles