missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Rectangular Fluorinated HDPE Carboy with Spigot

Easily fill and clean this carboy. Featuring a wide-mouth opening, space-saving rectangular design and a fluorinated surface to enhance long-term performance.

$430.19 - $447.52

Specifications

Closure Size 100mm
Description Fluorinated Rectangular Carboy w/Spigot
Diameter (Metric) Inner Neck 86mm
Material Fluorinated HDPE
Spigot Molded-in spigot boss
View More Specs

Alert:

Spigoted Carboys not for use with flammable liquids.

Disclaimers:

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.



Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Size Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Closure Size Capacity (Metric) Price Quantity  
02-961-57F Thermo Scientific™
NDS23270020
100mm 9L Each for $430.19
02-963-5D Thermo Scientific™
NDS23270050
100mm 20L Case for $447.52
Specifications

Description

Easily fill and clean the Thermo Scientific™ Nalgene™ Rectangular Fluorinated HDPE Carboy with Spigot, which features a wide-mouth opening and a sturdy stainless-steel handle. These Carboys are ideal for storing and dispensing a variety of solvents, and the fluorinated surface (inside and out) improves barrier properties and reduces solvent absorption while enhancing long-term container performance to prevent loss due to permeation.

Features:
  • Molded-in graduations of 1 gallon and 5L increments
  • Molded-in spigot boss provides strength and leak resistance
  • Rectangular shape saves valuable space
  • Graduated

Includes:

ETFE spigot and fluorinated white polypropylene screw closure

Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com

Description & Specifications

Specifications

100mm
86mm
Molded-in spigot boss
Fluorinated Rectangular Carboy w/Spigot
Fluorinated HDPE