Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with SFCA Membrane
Perform vacuum filtration and single-use biological work requiring sterile filtration, prefiltration or clarification of aqueous solutions in the lab or field.
$71.29 - $960.94
Specifications
Membrane SFCA
No. per Pack 12
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Diameter (Metric) Membrane Pore Size Volume (Metric) Upper Price Quantity    
09-740-36M
thermo scientific™
N1220020
50mm 0.2μm 115mL Pack of 12 Each for $292.70
Case of 6 Pack for $1,756.20
09-740-36N
thermo scientific™
N1220045
50mm 0.45μm 115mL Pack of 12 Each for $292.70
Case of 6 Pack for $1,756.20
09-740-28E
thermo scientific™
N1550020
50mm 0.2μm 150mL Case of 12 Each for $335.82
09-740-28F
thermo scientific™
N1550045
50mm 0.45μm 150mL Case of 12 Each for $335.82
09-740-28C
thermo scientific™
N1564020
75mm 0.2μm 500mL Case of 12 Each for $606.94
09-740-28D
thermo scientific™
N1564045
75mm 0.45μm 500mL Case of 12 Each for $606.94
09-740-39A
thermo scientific™
N1570020
50mm 0.2μm 250mL Case of 12 Each for $360.45
09-740-39B
thermo scientific™
N1570045
50mm 0.45μm 250mL Case of 12 Each for $360.45
09-740-40A
thermo scientific™
N1580020
75mm 0.2μm 1000mL Case of 12 Each for $828.76
09-740-40B
thermo scientific™
N1580045
50mm 0.45μm 1000mL Case of 12 Each for $828.76
09-740-43A
thermo scientific™
N1610020
90mm 0.2μm 1000mL Case of 12 Each for $960.94
09-740-43B
thermo scientific™
N1610045
90mm 0.45μm 1000mL Case of 12 Each for $960.94
09-740-44A
thermo scientific™
N1620020
90mm 0.2μm 500mL Case of 12 Each for $790.39
09-740-44B
thermo scientific™
N1620045
90mm 0.45μm 500mL Each for $71.29
Case of 12 Each for $855.48
Description & Specifications
Specifications
Membrane SFCA
No. per Pack 12
Conduct vacuum filtration and single-use biological work requiring sterile filtration, prefiltration or clarification of aqueous solutions in the lab or field with Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with SFCA Membrane. Surfactant-Free Cellulose Acetate (SFCA) membranes have lower extractables than standard (CA) membranes.
Features
  • Upper chamber separates from storage bottle for easy access to filtrate
  • Upper chamber clearly marked with pore size, membrane type, catalog number, lot number and expiration date (except 115 mL size)
  • Leakproof† (except 115 mL sizes) screw cap eliminates pH shift in storage bottle to provide longer life for stored media
  • Padless membrane support minimizes foaming of proteinaceous samples
  • Non-pyrogenic and non-cytotoxic
  • Gamma radiation-sterilized and individually bagged for a 5-year sterile shelf life
Surfactant-Free Cellulose Acetate Membranes (SFCA)
  • Low protein binding, excellent flow rates, little or no loss of specific proteins
  • No wetting agents to affect sensitive cell culture lines
  • Yellow collar for easy identification 
  • Much lower extractables than standard CA membranes 
115mL Sizes
  • Compact filter units for quick filtration of 50 to 100mL of fluid
  • Unitary construction with non-separable upper and lower compartments
  • Not intended for post-filtration storage of media (use Rapid-Flow products if media storage is required)
  • 115mL sizes do not have Rapid-Flow support plate design
Includes:
Polystyrene cover; graduated upper chamber and storage bottle (except 115mL size) integral membrane filter; cellulosic-plugged side arm; quick disconnect tubing adapter.
Warranty: 90 days
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.