Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with Nylon Membrane
Filter solutions where very low extractables are desired and solutions containing weak solvents with these disposable filter units with nylon membrane.
$85.58 - $1,183.90
Specifications
Membrane Nylon
No. per Case 12
No. per Pack 12
View More Specs
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Membrane Pore Size Volume (Metric) Upper Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Membrane Pore Size Volume (Metric) Upper Price Quantity  
09-740-31F Thermo Scientific™
N1500020
50mm 0.2μm 150mL Case of 12 Each for $460.33
09-740-31G Thermo Scientific™
N1500045
50mm 0.45μm 150mL Case of 12 Each for $419.79
09-740-32 Thermo Scientific™
1514020
75mm 0.2μm 500mL Case of 12 Each for $745.61
09-740-32B Thermo Scientific™
N1514045
75mm 0.45μm 500mL Case of 12 Each for $745.61
09-740-24A Thermo Scientific™
N1530020
50mm 0.2μm 250mL Case of 12 Each for $482.45
09-740-24B Thermo Scientific™
N1530045
50mm 0.45μm 250mL Case of 12 Each for $482.45
09-740-26A Thermo Scientific™
N1540020
75mm 0.2μm 1000mL Case of 12 Each for $921.06
09-740-26B Thermo Scientific™
N1540045
75mm 0.45μm 1000mL Case of 12 Each for $921.06
09-740-45 Thermo Scientific™
1630020
90mm 0.2μm 500mL Each for $85.58
Case of 12 Each for $1,026.96
09-740-46 Thermo Scientific™
1640020
90mm 0.2μm 1000mL Case of 12 Each for $1,183.90
Specifications
Description
Filter solutions where very low extractables are desired and solutions containing weak solvents such as alcohols with Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with Nylon Membrane.
Upper Chamber
  • Separates from receiver for easy access to filtrate
  • Clearly marked with pore size, membrane type, catalog number, lot number and expiration date (except 115mL size) for traceability
Receiver Bottle
  • Leakproof† screw cap eliminates pH shift in receiver to ensure longer life of stored fluids
  • An ideal filtrate storage vessel
Nylon Membrane
  • Low extractables and naturally hydrophilic
  • Contains no wetting agents and is alcohol resistant
  • Red collar for easy identification of correct membrane for your application
Other Features
  • Padless membrane support minimizes foaming of proteinaceous samples
  • Non-pyrogenic and non-cytotoxic
  • Gamma radiation sterilized and individually bagged for a 5-year sterile shelf life
Includes:
Polystyrene cover; graduated upper chamber and storage bottle; integral membrane filter; cellulosic-plugged side arm; quick-disconnect tubing adapter.
Note:
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Warranty: 90 days
Description & Specifications
Specifications
Nylon
12
12