Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with CN Membrane
Perform sterile filtration, prefiltration or clarification of buffers and other aqueous solutions when protein binding is not a concern with these filter units.
$331.18 - $1,009.70
Specifications
Membrane Cellulose Nitrate
No. per Pack 12
View More Specs
Products 14
Catalog Number Mfr. No. Volume (Metric) Upper Pore Size Volume (Metric) Receiver Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Upper Pore Size Volume (Metric) Receiver Price Quantity  
09-740-36K View Documents Thermo Scientific™
N1210020
115mL 0.2μm 115mL Pack of 12 Each for $331.18
Case of 6 Pack for $1,987.08
09-740-36L View Documents Thermo Scientific™
N1210045
115mL 0.45μm 115mL Pack of 12 Each for $331.18
Case of 6 Pack for $1,987.08
09-740-1A View Documents Thermo Scientific™
N1250020
150mL 0.2μm 150mL Case of 12 Each for $383.39
09-740-1B View Documents Thermo Scientific™
N1250045
150mL 0.45μm 150mL Case of 12 Each for $383.39
09-740-1C View Documents Thermo Scientific™
N1250080
150mL 0.8μm 150mL Case of 12 Each for $383.39
09-740-2A View Documents Thermo Scientific™
N1260020
250mL 0.2μm 250mL Case of 12 Each for $413.82
09-740-2B View Documents Thermo Scientific™
N1260045
250mL 0.45μm 250mL Case of 12 Each for $413.82
09-740-2C View Documents Thermo Scientific™
N1260080
250mL 0.8μm 250mL Case of 12 Each for $448.64
09-740-25A View Documents Thermo Scientific™
N4500020
500mL 0.2μm 500mL Case of 12 Each for $690.73
09-740-25B View Documents Thermo Scientific™
N4500045
500mL 0.45μm 500mL Case of 12 Each for $690.73
09-740-25AA View Documents Thermo Scientific™
N4500080
500mL 0.8μm 500mL Case of 12 Each for $690.09
09-740-3A View Documents Thermo Scientific™
N1270020
500mL 0.2μm 1000mL Case of 12 Each for $937.21
09-740-3B View Documents Thermo Scientific™
N1270045
500mL 0.45μm 1000mL Case of 12 Each for $937.21
09-740-3C View Documents Thermo Scientific™
N1270080
500mL 0.8μm 1000mL Case of 12 Each for $1,009.70
Description
Description
Use Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with CN (Cellulose Nitrate) membrane for sterile filtration, prefiltration or clarification of buffers and other aqueous solutions when protein binding is not a concern.
CN Membrane
  • Triton™-free
  • Has a green collar for easy identification of correct membrane type
Upper Chamber
  • Separates from receiver for easy access to filtrate
  • Clearly marked with pore size, membrane type, catalog number, lot number and expiration date for traceablility
Receiver Bottle
  • Leakproof† screw cap eliminates pH shift in receiver bottle to provide longer life for stored fluids
  • An ideal filtrate storage vessel
Other Features
  • Padless membrane support minimizes foaming of proteinaceous samples
  • Non-pyrogenic and non-cytogenic
  • Gamma radiation-sterilized and individually wrapped for a five-year sterile shelf life
115mL Sizes
  • Compact filter units for quick filtration of 50 to 100mL of fluid
  • Unitary construction with non-separable upper and receiver components
  • Side arm on reciever 
  • Not intended for post-filtration storage of media (use Rapid-Flow products if media storage is required)
  • 115mL sizes do not have Rapid-Flow support plate design
Includes:
50 or 75mm diameter membrane, polyethylene adapter, and quick-disconnect tubing adapter. Each pack of 500 and 1000mL units includes 12 glass fiber prefilters. All sizes except 115mL supplied with sterile polyethylene leakproof closure that lets you use the receiver for filtrate storage.
Note:
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Warranty: 90 days
Specifications
Specifications
Cellulose Nitrate
12
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Note: CN membranes are best used for filtration of buffers and general lab solutions. We do not recommend CN for filtration of cell culture fluids because it has high protein binding characteristics. Use PES or SFCA for cell culture media, serum, additives, etc.