Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with SFCA Membrane
Lower the number of extractables in your bottle-top filters by switching from standard CA membranes to sterile, disposable bottle top filters with SFCA Membrane.
$299.15 - $683.97
Specifications
No. per Case 12
No. per Pack 12
Sterility Sterile
View More Specs
Products 10
Catalog Number Mfr. No. Volume (Metric) Pore Size (Metric) Fits Bottle Neck Size (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Pore Size (Metric) Fits Bottle Neck Size (Metric) Price Quantity  
09-740-22D View Documents Thermo Scientific™
N2903320
150 mL 0.2 μm 33 mm Case of 12 Each for $299.15
09-740-22E View Documents Thermo Scientific™
N2903345
150 mL 0.45 μm 33 mm Case of 12 Each for $305.97
09-740-22F Accessories View Documents Thermo Scientific™
N2904520
150 mL 0.2 μm 45 mm Case of 12 Each for $299.15
09-740-22G Accessories View Documents Thermo Scientific™
N2904545
150 mL 0.45 μm 45 mm Case of 12 Each for $305.91
09-740-22H View Documents Thermo Scientific™
N2913320
500 mL 0.2 μm 33 mm Case of 12 Each for $416.48
09-740-22J View Documents Thermo Scientific™
N2913345
500 mL 0.45 μm 33 mm Case of 12 Each for $416.97
09-740-22K Accessories View Documents Thermo Scientific™
N2914520
500 mL 0.2 μm 45 mm Case of 12 Each for $416.48
09-740-22L Accessories View Documents Thermo Scientific™
N2914545
500 mL 0.45 μm 45 mm Case of 12 Each for $416.48
09-740-47A Accessories View Documents Thermo Scientific™
N2923320
1000 mL 0.2 μm 33 mm Case of 12 Each for $683.97
09-740-47C View Documents Thermo Scientific™
N2924520
1000 mL 0.2 μm 45 mm Case of 12 Each for $662.73
Description
Description
Lower the number of extractables in your bottle-top filters by switching from standard cellulose acetate (CA) to Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with SFCA Membrane. These filters fit Nalgene filter receiver bottles (#455-series) or standard glass media bottles and have surfactant-free cellulose acetate (SFCA) membranes.
Bottle Top Filters:
  • Marked with catalog number, membrane type, pore size, lot number and expiration date for easy traceability
  • Gamma radiation sterilized and individually bagged for 5-year sterile shelf life
  • Molded-in graduations for quick visualization of sample volume
  • Side-arm with quick-disconnect tubing adapter for easy bottle top changes
  • Non-cytotoxic and non-pyrogenic
Surfactant-Free Cellulose Acetate Membranes (SFCA)
  • Low protein binding with little or no loss of specific proteins
  • Excellent flow rates
  • No wetting agents to affect sensitive cell culture lines
  • Much lower extractables than standard CA membranes 
Includes:
Polystyrene graduated chamber; color-coded collar with cellulosic vent plug side arm and quick-disconnect tubing adapter; 50, 75, or 90mm diameter membranes; bottom screw threads for attachment to bottles with 33 or 45mm neck sizes.
Warranty: 90 days
Specifications
Specifications
12
12
Sterile
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.