missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Achieve fast flow rates and high throughput for increased filtration efficiency with Nalgene Rapid-Flow sterile disposable bottle top filters.
Supplier: Thermo Scientific™ 2904520
Alert:
CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.
Description
Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.
- Available in PES and SFCA membranes. Devices are color coded according to membrane.
- Devices feature the fastest flow rate and lowest clogging of comparable alternatives
- 0.1um PES available for mycoplasma protection
- 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
- Non-pyrogenic and endotoxin free - Rapid-Flow filters meet the most rigorous testing standards for any vacuum filtration unit
- Available with Fit for Purpose Sterile documentation
Specifications
Single-use | |
Polystyrene | |
0.2 μm | |
45mm | |
12 | |
150 mL | |
50 mm |
Membrane Filter | |
SFCA | |
Sterile | |
12 | |
Bottle Top Filter | |
50 mm | |
Sterile |