Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Bottle, 175mL
Perform pelleting applications such as large-volume cell harvesting, pelleting and protein purification with these PPCO conical-bottom centrifuge bottles.
$2,204.12 - $2,204.12
Products
Catalog Number Mfr. No. Max. Force Nominal Capacity Price Quantity    
05-579-28
thermo scientific™
3143-0175
27,500 x G 175mL Case of 36 Each for $2,204.12
Description & Specifications
Perform pelleting applications such as large-volume cell harvesting, pelleting and protein purification with Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Bottles, which are molded of rigid and robust polypropylene copolymer. These translucent bottles are autoclavable and have excellent chemical resistance properties.
Features:
  • For low to moderate speed centrifugation of biological and chemical samples
  • For proper performance, bottles must be filled to 80% of the total capacity
  • Must be used with Nalgene adapter for conical-bottom centrifuge bottles (DS3126-0175)
  • Autoclavable, graduated and leakproof†
Includes:
Bottle includes polypropylene sealing cap assembly with silicone gasket for added leakproof assurance at maximum speeds or when spinning hazardous materials
Note: These tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.