Thermo Scientific™ Nalgene™ Polysulfone Filter Holder with Funnel
Use the Thermo Scientific™ Nalgene™ Polysulfone Filter Holder with Funnel with any filtering flask or manifold that accepts a suitable rubber stopper with hole.
$321.06 - $474.27
Specifications
Autoclavable Yes
Material Polysulfone
View More Specs
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
09-745 Thermo Scientific™
NDS03104000
Each for $321.06
09-746 Thermo Scientific™
DS03104050
Each for $474.27
Specifications
Description
Use the Thermo Scientific™ Nalgene™ Polysulfone Filter Holder with Funnel in conjunction with any filtering flask or manifold that accepts a suitable rubber stopper with hole. These filter holders with funnel provide an environmentally friendly alternative to disposable filters and are made of washable, autoclavable polysulfone.
Features:
  • Reusable, autoclavable polysulfone lasts for years
  • Graduated upper chamber for easy sample measurement
  • Includes analytical and sterilization support plates
  • Use with filtering flasks or manifolds
Includes:
  • Rugged polysulfone funnel
  • Polypropylene copolymer cover with three ports
  • Three friction-fit TPE caps
  • Support plate for 47mm membrane
  • No-twist vacuum silicone gasket which can replace a No. 7 or 8 rubber stopper with 0.56″ (15mm) hole
  • Aluminum clamp
Warranty: 90 days
Description & Specifications
Specifications
Yes
Polysulfone