Thermo Scientific™ Nalgene™ Polyproylene Pneumatic Trough
Replace metal-type troughs with the lightweight, corrosion-proof and quiet Thermo Scientific™ Nalgene™ Polypropylene Pneumatic Trough.
$89.48 - $89.48
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
15-310 Thermo Scientific™
N68200010
Polypropylene Pneumatic Trough Each for $89.48
Case of 6 Each for $536.88
Specifications
Description
Replace metal-type troughs with the lightweight, corrosion-proof and quiet Thermo Scientific™ Nalgene™ Polypropylene Pneumatic Trough. This trough features tapered sides for nesting and a built-in gas inlet. There are no sharp corners or separate parts to lose. The built-in platform accommodates four 250mL bottles.
  • Lightweight and rustproof
  • Gas tubing passes through a loop to prevent liquid from flowing into gas line
  • Liquid inlet and outlet connections accept 1/4" I.D. × 3/8″ O.D. (6.4 × 9.5mm) tubing (tubing sold separately)
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.