Thermo Scientific™ Nalgene™ Polypropylene Stirring Rod
Combine functions of a stirring rod and rubber policeman with these polypropylene stirring rods, featuring a flat paddle at one end and a triangular paddle at the other end.
$109.52 - $109.52
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
14-518 Thermo Scientific™
N61690010
Pack of 12 Each for $109.52
Case of 6 Pack for $657.12
Specifications
Description
Combine the functions of a stirring rod and rubber policeman with Thermo Scientific™ Nalgene™ Polypropylene Stirring Rods, featuring a flat paddle at one end and a triangular paddle at the other end. These autoclavable stirring rods are chemically compatible with a wide range of lab reagents.
  • Flat paddle, 5/8 x 5/8″ (1.6 x 1.6cm) at one end; triangular paddle at the other end
  • Overall length is 9.75″ (24.8cm), makes it a convenient and conventional length for a wide variety of lab uses
  • Usable over a temperature range of 0°C to +135°C making it suitable for use in heated applications
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.