Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps
Avoid corrosion from acid exposure with Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps, which are chemically compatible with most lab reagents.
$81.32 - $81.32
Products
Catalog Number Mfr. No. Price Quantity    
10-309
thermo scientific™
N63200010
Pack of 12 Each for $81.32
Case of 6 Pack for $487.92
Description & Specifications
Avoid corrosion from acid exposure with Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps, which are chemically compatible with most lab reagents. Double-action hinges provide extra leverage and long life. These forceps feature a scissor-type handle with a ratchet to lock tips shut and a handy clamp for chromatography columns.
  • Autoclavable
  • Feature jaws with interlocking teeth to grip objects tightly
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.