Thermo Scientific™ Nalgene™ Polypropylene Replacement Coupling Inserts for Quick Filling/Venting Closures
Promote faster installation using these replacement coupling inserts, which support one-handed connection and disconnection when applied to mating coupling bodies.
$68.30 - $324.61
Specifications
Material Polypropylene
No. per Case 6
No. per Pack 6
View More Specs
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Diameter (English) Inner Tubing Price Quantity  
Catalog Number Mfr. No. Description Diameter (English) Inner Tubing Price Quantity  
02-923-14 Thermo Scientific™
21590001
Straight-through coupling insert 1/4 in. Case of 6 Each for $68.30
02-923-16 Thermo Scientific™
2159002
Straight-through coupling insert 3/8 in. Case of 6 Each for $72.72
02-923-18 Thermo Scientific™
21590012
90° Elbow coupling insert 3/8 in. Case of 6 Each for $324.61
02-923-17 Thermo Scientific™
21590011
90° Elbow coupling insert 1/4 in. Case for $298.36
Specifications
Description
Promote faster installation using Thermo Scientific™ Nalgene™ Polypropylene Replacement Coupling Inserts for Quick/Filling Venting Closures, which support one-handed connection and disconnection when applied to mating coupling bodies. Male, barbed quick-disconnect replacement coupling inserts for use with Nalgene Quick Filling/Venting Closures.
  • For use with Nalgene Quick Filling/Venting Closures
  • Promotes fewer leakpoints and short assemblies
Warranty: 90 days
Description & Specifications
Specifications
Polypropylene
6
6