Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Polypropylene Quick Action Spigots
Incorporate Thermo Scientific™ Nalgene™ Polypropylene Quick-Action Spigots, including an HDPE tubing adapter which accepts tubing with 1/4 or 5/16″ I.D with Carboys.
$83.55 - $83.55
Products
Catalog Number Mfr. No. Description Price Quantity    
02-963-7
thermo scientific™
64220010
Polypropylene Quick Action Spigots Each for $83.55
Case of 12 Each for $1,002.60
Description & Specifications
Incorporate these Thermo Scientific™ Nalgene™ Polypropylene Quick-Action Spigots, including an HDPE tubing adapter which accepts tubing with 1/4 or 5/16″ I.D with Nalgene Carboys to increase functionality.
Features:
  • Take 5/8″ (1.6cm) I.D. tubing
  • Supplied with HDPE adapter for 1/4 or 5/16″ (0.6 or 0.8cm) tubing
Notes:
  • Disassemble spigot before autoclaving; tubing adapter is not autoclavable
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.