Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
Reduce breakage and chipping of glass pipette tips with the stackable and lightweight Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray.
$158.53 - $158.53
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
13-361-5 Thermo Scientific™
N69100618
Sterilizing tray with rigid plastic construction Each for $158.53
Case of 6 Each for $951.18
Specifications
Description
Reduce breakage and chipping of glass pipette tips with the Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray. Horizontal design allows pipettes to be fully immersed in disinfectant. Rigid plastic tray is stackable, lightweight, quiet and less expensive than stainless steel. Cover fits into groove of pan to minimize spillage of hazardous materials.
  • Holds pipets up to 40cm long in a stackable, lightweight alternative to stainless steel
  • Can also be used to sterilize small labware and instruments—resists aggressive disinfectants
  • Has groove for cover to minimize spillage of hazardous materials and a hand grip on the cover
  • Autoclavable
Compliance:
  • Meets OSHA Standard 29CFR1910.1030 for use as protection against bloodborne pathogens.
  • Conforms to NIH and Federal Register guidelines for biohazardous and recombinant DNA research where disinfecting and subsequent autoclaving of pipets are required before cleaning and disposal
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.