Thermo Scientific™ Nalgene™ Polypropylene Heavy-Duty Vacuum Carboys
Ideal for the most extreme service conditions, this heavy-duty vacuum carboy is useful as a vacuum trap and will hold full vacuum up to eight hours.
$382.26 - $578.19
Specifications
Description Heavy-Duty Vacuum Carboys
Autoclavable Yes
Closure Size 83B
Handles Yes
Material Polypropylene
View More Specs
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
02-960-14 Thermo Scientific™
22260020
10L 83B Pack for $382.26
Case of 6 Pack for $1,994.40
02-960-15 Thermo Scientific™
22260050
20L 83B Pack for $578.19
Case of 4 Pack for $2,011.08
Specifications
Description
Ideal for the most extreme service conditions, the Thermo Scientific™ Nalgene™ Polypropylene Heavy Duty Vacuum Carboy is useful as a vacuum trap and will hold a full vacuum up to eight hours. Constructed of thick-walled natural polypropylene to provide added strength, it is supplied with white 83B PP closure with TPE (thermoplastic elastomer) gasket. Customization options available for fluid transfer applications. Fit perfectly with a variety of optional 83B closures with standard tubing sets (such as Nalgene™ Top Works™, Nalgene™ Filling/Venting) and custom tubing set designs.
  • Autoclavable when closure threads are disengaged and vessel is empty
  • Not rated for pressure use
Ordering Information:
  • For closure options in vacuum applications, use with Nalgene Quick Filling/Venting Closures (Cat. No. 2158), or Nalgene Filling/Venting Closures (Cat. No. 2162)
  • Additional accessories include Nalgene Barbed Bulkhead Fittings (Cat. No. 6149)
  • For 1 to 5L sizes, please refer to Nalgene Heavy Duty Vacuum Bottles (Cat. No. 2126)
Warranty: 90 days
Description & Specifications
Specifications
Heavy-Duty Vacuum Carboys
83B
Polypropylene
Yes
Yes
1