Thermo Scientific™ Nalgene™ Polypropylene End Caps
Securely close off 3/4″ or 3″ sanitary ports on carboys with Thermo Scientific™ Nalgene™ Polypropylene End Caps.
$192.52 - $192.52
Specifications
No. per Case 1
No. per Pack 1
View More Specs
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
12-565-976 Thermo Scientific™
N26650300
3 in. Tri (76mm) Each for $192.52
Specifications
Description
Securely close off 3/4″ or 3″ sanitary ports on carboys with Thermo Scientific™ Nalgene™ Polypropylene End Caps. These are reliable, repeatable seals when used in combination with a silicone gasket and true union clamp.  A groove on underside of end cap accepts standard sanitary gaskets for sealing the systems.
  • Seals sanitary ports on Nalgene Sanitary Carboys (No. 2630 and 2640)
  • USP Class VI
  • Autoclavable
Warranty: 90 days
Description & Specifications
Specifications
1
1