Thermo Scientific™ Nalgene™ Polypropylene Desiccator with Stopcock
Clearly view contents with the transparent PC cover of this polypropylene desiccator. Lighter in weight than glass, it is compatible with all solid desiccants.
$253.44 - $253.44
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
08-642-5 Accessories Thermo Scientific™
N53100250
Each for $253.44
Case of 4 Each for $1,013.76
Specifications
Description
Clearly view contents with the transparent PC cover of the Thermo Scientific™ Nalgene™ Polypropylene Desiccator with Stopcock. Lighter in weight than glass, the robust polypropylene body is compatible with all solid desiccants and protects against breakage. This desiccator is for room temperature use only and can be used with or without a vacuum.
Features:
  • Full vacuum may be applied following supplied instructions for proper performance
  • Polypropylene stopcock with PTFE Plug can be used with 0.25″ (6.4mm) ID tubing (tubing sold separately)
  • Vacuum connection is located on the desiccator body making the cover easier to handle
  • Straight bore design eliminates stopcock vacuum leakage
  • No gasket is necessary; grease makes polycarbonate cover airtight even without applying a vacuum
  • Recessed rim keeps grease off benchtop when cover is set down
  • Desiccator takes any standard 230mm desiccator plate such as Cat. No. 5312-0230 (sold separately)
Note: Desiccator is not autoclavable
Includes:
Cover, Stopcock
Requires:
230mm plate such as Nalgene 5312-0230. (metal plate pictured not included and not available)
Ordering Information:
Replacement cover available (Cat. No. 715311-0250)
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.