Thermo Scientific™ Nalgene™ Polycarbonate Validation Bottles
Perform QA lot sample storage of high-valued pharmaceuticals using Nalgene Polycarbonate Validation Bottles. Small-volume bottles require lower volume of expensive reagents.
$310.14 - $1,194.37
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
02-923-10 Thermo Scientific™
DS21270250
250mL 53Bmm Case of 6 Each for $310.14
36-103-1180 Thermo Scientific™
DS21272000
2L 53Bmm Case of 12 Each for $1,018.80
36-104-0070 Thermo Scientific™
DS21270030
30mL 20-415mm Case of 30 Each for $1,194.37
Specifications
Description
Perform QA lot sample storage of high-valued pharmaceuticals using Thermo Scientific™ Nalgene™ Polycarbonate Validation Bottles. Small-volume bottles are molded of the same materials as larger carboys, but require a lower volume of expensive reagents to perform validation testing.
Autoclavable, transparent polycarbonate construction allows for convenient material compatibility validation.
Includes:
White polypropylene screw closure and TPE gasket
Warranty: 90 days