Thermo Scientific™ Nalgene™ Polycarbonate Safety Splash Shield
Simultaneously maintain safety and a clear field of vision by using this safety splash shield. Provides protection from splashes, aerosols and flying debris.
$472.61 - $472.61
Alert:
Warning!: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
14-293-38 Thermo Scientific™
N63560001
Safety Splash Shield with acetal flexible arm; PC 21.6 x 27.9 cm Each for $472.61
Case of 4 Each for $1,890.44
Specifications
Description
Simultaneously maintain safety and a clear field of vision by using the Thermo Scientific™ Nalgene™ Polycarbonate Safety Splash Shield. This shield attaches easily to benchtop shelving, machinery or other magnetic surfaces for convenient protection from splashes, aerosols and flying debris. This durable, scratch-resistant 1/8″ (3mm) thick polycarbonate shield protects the user and features an easy to maneuver arm. Meets OSHA 29CFR1910.1030 standard.
  • Flexible 14 inch arm makes it easy to maneuver shield over samples
  • Magnetic base firmly grips to magnetic surfaces letting you position the shield horizontally (flat), vertically or on an overhead surface
  • Provides UV protection from 200-360nm for added versatility
  • Biohazard/Transparent
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.