Thermo Scientific™ Nalgene™ Polycarbonate Imhoff Settling Cone and Rack
Determine settleable matter in water and waste effluent and measure the sand content in well water with this settling cone, made of durable polycarbonate with screw closure.
$106.45 - $106.45
Disclaimers:
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Note: These tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
15-438 Thermo Scientific™
N10000010
Settling Cone Each for $106.45
Case of 4 Each for $425.80
Specifications
Description
Determine settleable matter in water and waste effluent and also measure the sand content in well water. The Thermo Scientific™ Nalgene™ Polycarbonate Imhoff Settling Cone and Rack are constructed from durable polycarbonate. The cone features a polypropylene screw closure. The leakproof† closure makes it possible to withdraw sediment for gravimentric anaylsis, and also aids in cleaning. Closure design enables user to read graduations to zero.
Features:
  • Designed for determining settleable matter in water and waste effluent or sand content in well water
  • Closure design enables user to read graduations to zero
  • Autoclavable/Transparent/Graduated/Leakproof
  • Graduated from 0 to 1000mL, with 0.1mL increments in the 0 to 1mL range
  • 13mm leakproof† screw closure at bottom for withdrawing sediments
Settling Cone Graduation Intervals
  • 0-1mL: 0.1mL
  • 1-10mL: 0.5mL
  • 10-20mL: 1mL
  • 20-40mL: 2mL
  • 40-100mL: 10mL
  • 100-500mL: 50mL
  • 500-1000mL: 100mL
Ordering Information:
Use with Imhoff cone rack Cat. No. 1001-0010 (sold separately).
<.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.