Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Polycarbonate Fernbach Culture Flask
Increase aeration for better growth of culture in liquid media with the wide surface area of these Nalgene Fernbach culture flasks.
$224.59 - $224.59
Products
Catalog Number Mfr. No. Price Quantity    
10-041-19
thermo scientific™
N41052800
Each for $224.59
Case of 4 Each for $898.36
Description & Specifications
Increase aeration for better growth of culture in liquid media with the wide surface area of these Thermo Scientific™ Nalgene™ Polycarbonate Fernbach Culture Flask. Constructed of durable and transparent polycarbonate.
  • Break-resistant, transparent polycarbonate
  • Fernbach design supports better aeration
  • For leakproof seal† use No. 12 rubber stopper
Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.