Thermo Scientific™ Nalgene™ Polycarbonate Culture Vessel with Ports
Gain easy access to your suspension cultures with four-port, Nalgene™ Polycarbonate Culture Vessels. They are economical, lightweight and break-resistant.
$797.00 - $797.00
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
03-409-9H Thermo Scientific™
N26000012
12000mL Each for $797.00
Case of 2 Each for $1,594.00
Specifications
Description
Gain easy access to your suspension cultures with four-port, Thermo Scientific™ Nalgene™ Polycarbonate Culture Vessels with Ports. Economical, lightweight and break-resistant, it is molded of transparent polycarbonate for easy viewing of your cultures.
  • Graduated in 0.5L increments from 3 to 12L
  • For use with overhead drive mixers and lower assemblies for top to bottom mixing
  • Port accessories are available, ports accept any Nalgene 38-430 size closure
  • Noncytotoxic
  • Autoclavable and USP Class VI compliant
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.