Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Polycarbonate Conical-Bottom Centrifuge Bottle
Process pelleting applications such as cell culture harvesting and protein biochemistry with this polycarbonate conical-bottom centrifuge bottle.
$2,363.55 - $2,363.55
Products
Catalog Number Mfr. No. Max. Force Nominal Capacity Price Quantity    
05-579-29
thermo scientific™
31440175
27,500 x G 175mL Case of 36 Each for $2,363.55
Description & Specifications
Process pelleting applications such as cell culture harvesting and protein biochemistry, including ammonium sulfate precipitations, with Thermo Scientific™ Nalgene™ Polycarbonate Conical-Bottom Centrifuge Bottle. Molded of rigid and robust polycarbonate copolymer, these transparent bottles are autoclavable and offer excellent mechanical strength.
Features:
  • For low to moderate speed centrifugation of biological and chemical samples
  • Includes polypropylene sealing cap assembly with silicone gasket for added leakproof† assurance at maximum speeds or when spinning hazardous materials
  • Must be used with Nalgene adapter for conical-bottom centrifuge bottles (DS3126-0175)
  • For proper performance, bottles must be filled to 80% of total capacity
  • Autoclavable, transparent, leakproof† with molded-in graduations
Note: These tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.