missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing for Peristaltic Pumps

Ensure consistently reliable and accurate fluid transfers. This durable, high-purity tubing is ideal for pharmaceutical, laboratory and bioprocess manufacturing.

Manufacturer:  Thermo Scientific™ 86003018

 View more versions of this product

Catalog No. 14-179-128



Description

Description

Ensure reliable and accurate fluid transfers every time with Thermo Scientific™ Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing. This flexible, durable high-purity translucent tubing is designed for a variety of transfer applications, including pharmaceutical, laboratory and bioprocess manufacturing.

  • Tasteless, odorless, non-cytotoxic, non-pyrogenic and non-hemolytic
  • Contains no phthalate plasticizers or peroxides that can leach out into transported fluid
  • Low binding surface minimizes absorption of proteins and food products
  • Autoclavable; also can be sterilized by gamma radiation, EtO and chemical procedures
  • Durometer hardness: Shore A, 50
  • Meets FDA, USDA, U.S. Pharmacopeia Class VI, 24th Edition; and 3A sanitary standards

Ordering Information

Double-bagged in case of 25″ (7.6m)

Note:

Not intended for invasive use.

Specifications

Specifications

5/16 in.
25 ft.
No. 18
Durometer 50(A)
Silicone (Platinum-Cured)
Flexible Tubing
Translucent
Bend Radius 4xOD
SDS
Documents
Product Certifications
Promotions

Promotions

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.