Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ One-Piece Disposable LDPE Droppers
Manufacture 30 to 35 drops per mL with the bellows-type bulb of the Thermo Scientific™ Nalgene™ One-Piece Disposable LDPE Dropper.
$129.79 - $129.79
Products
Catalog Number Mfr. No. Description Price Quantity    
13-711-10
thermo scientific™
N62190068
Pack of 100 Each for $129.79
Case of 10 Pack for $1,297.90
Description & Specifications
Deliver 30 to 35 drops per mL with the bellows-type bulb of the Thermo Scientific™ Nalgene™ One-Piece Disposable LDPE Dropper. Molded in one piece, the dropper bulb provides excellent drop formation. This low density polyethylene dropper is safer than glass droppers and economical, featuring a non-wetting tapered tip and visible liquid level.
  • Chemically compatible with most acids, bases and alcohols
  • Molded in one piece, the dropper and bulb provide excellent drop formation
  • Safe and economical for reliable performance
  • Nonwetting tapered tip delivers 30 to 35 drops per mL
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.