Thermo Scientific™ Nalgene™ Oak Ridge Polysulfone Centrifuge Tubes w/Sealing Caps, 50mL
Easily view contents using strong, transparent Thermo Scientific™ Nalgene™ Oak Ridge Polysulfone Centrifuge Tubes, featuring an enhanced resistance to acids and bases.
$522.35 - $522.35
Alert:
Must be filled to at least 80% capacity for proper performance
Disclaimers:
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
05-563-5G Thermo Scientific™
N31370050
Pack of 10 Each for $522.35
Case of 5 Pack for $2,611.75
Specifications
Description
Easily view contents using strong, transparent Thermo Scientific™ Nalgene™ Oak Ridge Polysulfone Centrifuge Tubes, featuring an enhanced resistance to acids and bases.
Features:
  • Round bottoms fit high speed rotors
  • Transparent, with straw-colored cast
  • Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges
  • Autoclavable
  • Leakproof†
  • 3137-0050 comes with a PP sealing cap closure with silicone gasket for extra leak protection when spinning hazardous substances at >10,000 x g
Ordering Information:
Sealing cap assembly available separately (DS3132-series for 50mL sizes). Cat. No. 3137-0050 is designed to fit standard rotors (actual brim capacity is 42mL).Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.