Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Narrow-Mouth Translucent Amber HDPE Packaging Bottles with Closure: Bulk Pack
Safely store, ship and pack light-sensitive liquids with these translucent amber narrow-mouth bottles, featuring robust construction and amber PP closures.
$393.44 - $1,822.06
Specifications
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure Type Screw closure (bulk packed, unassembled)
Closure Color Opaque amber
Closure Material Polypropylene (PP)
Description Narrow-Mouth Packaging Bottles
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products
Catalog Number Mfr. No. Closure Size Capacity (Metric) Price Quantity    
14-178-173
thermo scientific™
N3120840001
20-415mm 30mL Case of 2000 Each for $1,822.06
0292397
thermo scientific™
3120840002
20-415mm 60mL Case of 1000 Each for $1,731.94
02923123
thermo scientific™
3120840004
24-415mm 125mL Case of 500 Each for $988.63
02923124
thermo scientific
3120840008
24-415mm 250mL Case of 250 Each for $757.86
0292398
thermo scientific™
3120840016
28-415mm 500mL Case of 125 Each for $528.83
02-923-61
thermo scientific™
3120840032
38-430mm 1000mL Case of 50 Each for $393.44
Description & Specifications
Specifications
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure Type Screw closure (bulk packed, unassembled)
Closure Color Opaque amber
Closure Material Polypropylene (PP)
Description Narrow-Mouth Packaging Bottles
Color Amber Translucent
Material HDPE
Mouth Narrow
Shape Round
Safely store, ship and package light-sensitive liquids with these translucent amber high density polyethylene narrow-mouth bottles. Thermo Scientific™ Nalgene™ Narrow-Mouth Translucent Amber HDPE Packaging Bottles with Closure have a robust and leakproof† construction for packaging use and feature amber polypropylene closures.
Features:
  • Excellent impact and chemical resistance
  • Include opaque amber polypropylene closures
  • Translucent amber bottles provide light protection with the ability to see liquid level
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton.
Warranty: 90 days