Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Packaging Bottles with Closure: Bulk Pack
Store, ship and package liquids with these robust, narrow-mouth bottles that are constructed for packaging use and include polypropylene closures.
$411.65 - $2,176.16
Specifications
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Type Screw closure (bulk packed, unassembled)
Closure Color Natural, translucent
Closure Material Polypropylene (PP)
Description Narrow-Mouth Packaging Bottles
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
02-923-99
View Documents
Thermo Scientific™
3120870001
30 mL 20-415 mm Case of 1000 Each for $2,133.03
03-313-9B
View Documents
Thermo Scientific™
3120870002
60 mL 20-415 mm Case of 1000 Each for $2,176.16
02-923-125
View Documents
Thermo Scientific™
3120870004
125 mL 24-415 mm Case of 500 Each for $1,216.00
033139D
View Documents
Thermo Scientific™
3120870008
250 mL 24-415 mm Case of 250 Each for $1,043.53
06444
View Documents
Thermo Scientific™
3120870016
500 mL 28-415 mm Case of 125 Each for $658.31
14-178-172
View Documents
Thermo Scientific™
N3120870032
1000 mL 38-430 mm Case of 50 Each for $411.65
Description
Description
Store, ship and package liquids with Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Packaging Bottles with Closure. Featuring excellent clarity, impact and chemical resistance, these polypropylene copolymer bottles have a robust construction for packaging use and are featured with polypropylene closures.
Features:
  • Natural, translucent polypropylene copolymer
  • Polypropylene screw closures
  • Autoclavable
  • Leakproof†
Warranty: 90 days
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton.
Specifications
Specifications
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Natural, translucent
Narrow-Mouth Packaging Bottles
PPCO
Round
Screw closure (bulk packed, unassembled)
Polypropylene (PP)
Natural Translucent
Narrow
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.