Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles without Closure: Bulk Pack


Protect light-sensitive product with these opaque amber packaging bottles, which meet requirements of light-resistant containers per USP latest edition.

Manufacturer: thermo scientific™  N3620850002

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Catalog No. 03-313-86B

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Capacity (Metric) 60mL
No. per Pack 1000
Capacity (English) 2 oz.
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition
Closure Color Opaque amber
Closure Screw closure (bulk packed, unassembled)
Closure Material Polypropylene (PP)
Closure Size 20-415mm
Color Amber Opaque
Description Narrow-Mouth Packaging Bottles
Diameter (Metric) 37mm
Diameter (Metric) Inner Neck 14mm
Height (Metric) 85mm
Mouth Narrow
No. per Case 1000
Shape Round
Capacity 2 oz., 60mL
Material HDPE
Neck Finish 20mm

Protect valuable light-sensitive reagents with Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles. Packed without closures, you can match these bottles with various colored closures (Cat. No. 362150 series). These high density polyethylene bottles meet the requirements of light-resistant containers per USP latest edition.

  • Opaque amber HDPE Bottles without closures
  • Meet USP requirements of light-resistant containers
  • Closures available separately (Series 362150)
  • Leakproof† with appropriate Nalgene closure

Ordering Information:

Bottles are bulk packed without closures. Order colored closures separately (Cat. No. 362150). † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.