Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Bottles with Closure: Shrink-Wrapped Trays
Experience easy handling with Nalgene Opaque Amber HDPE Bottles, conveniently packaged in shrink-wrap trays ready to feed into filling lines for various reagents.
$820.03 - $2,526.06
Specifications
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition
Closure Type Screw closure (bulk packed, unassembled)
Closure Color Opaque amber
Closure Material Polypropylene (PP)
Description Narrow-Mouth Packaging Bottles
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Closure Size Capacity (Metric) Price Quantity    
02-923-85
thermo scientific™
3220850001
20-415mm 30mL Case of 1050 Each for $2,411.13
02-923-87
thermo scientific™
3220850002
20-415mm 60mL Case of 1050 Each for $2,526.06
02-930-102
thermo scientific™
N3220850004
24-415mm 125mL Case of 1050 Each for $820.03
Description & Specifications
Specifications
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition
Closure Type Screw closure (bulk packed, unassembled)
Closure Color Opaque amber
Closure Material Polypropylene (PP)
Description Narrow-Mouth Packaging Bottles
Color Amber Opaque
Material HDPE
Mouth Narrow
Shape Round
Experience easy handling with Thermo Scientific™ Nalgene Opaque Amber HDPE Bottles, conveniently packaged in shrink-wrap trays ready to feed into filling lines for various reagents. Bottles include polypropylene closures.
  • Opaque amber container provides protection for light-sensitive reagents
  • Bottles are packed in rigid trays, allowing for easy handling
  • Opaque amber HDPE with opaque amber polypropylene closures
  • Leakproof†
Ordering Information:
Bottles are packed in shrink-wrapped trays with closures packed in separate polybags within the same carton.
Warranty: 90 days