Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Narrow-Mouth LDPE Packaging Bottles with Closure: Bulk Pack
Safely store, ship and package liquids with Thermo Scientific™ Nalgene™ Narrow-Mouth LDPE Packaging Bottles with Closure.
$751.34 - $1,722.88
Specifications
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Screw closure (bulk packed, unassembled)
Closure Color Natural, translucent
Closure Material Polypropylene (PP)
Description Narrow-Mouth Packaging Bottles
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity    
14-178-171
thermo scientific™
N3120880001
30mL 20-415mm Case of 1000 Each for $1,722.88
02-923-63
thermo scientific™
3120880008
250mL 24-415mm Case of 250 Each for $751.34
Description & Specifications
Specifications
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Screw closure (bulk packed, unassembled)
Closure Color Natural, translucent
Closure Material Polypropylene (PP)
Description Narrow-Mouth Packaging Bottles
Color Natural Translucent
Material LDPE
Mouth Narrow
Shape Round
Safely store, ship and package liquids with bulk-packed Thermo Scientific™ Nalgene™ Narrow-Mouth LDPE Packaging Bottles with Closure. With a robust and durable construction for packaging use, these bottles feature a polypropylene narrow mouth closure for easy pouring. The natural, low density polyethylene provides excellent flexibility, impact resistance and clarity.
Features:
  • Includes natural, polypropylene closures
  • Ideal material for trace metal analysis because of LDPE low trace metal content
  • Leakproof†
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton.