Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Bottles with Closure: Shrink-Wrapped Trays


Easily handle these narrow-mouth HDPE bottles that are ready to feed into filling lines for various reagents, and conveniently packaged in shrink-wrap trays.

Manufacturer: thermo scientific™  N3220890008

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. † Thermo Scientific Nalgene assigns the term “leakproof” a) To bottle/closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) To bottle/closure systems of 100 or 120mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, Thermo Scientific Nalgene advises customers to test bottles and closures under conditions of their planned applications. Nalgene bottles and carboys are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Catalog No. 02-930-103

  • / Case of 300 Each
 

Description & Specifications

Specifications

Capacity (English) 8 oz.
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Screw closure (bulk packed, unassembled)
Description Narrow-Mouth Packaging Bottles with Closures
Diameter (Metric) Inner Neck 18mm
Neck Finish 24mm
No. per Case 300
Shape Round
Capacity (Metric) 250mL
Closure Color Natural
Closure Material Polypropylene (PP)
Closure Size 24-415mm
Color Natural
Diameter (Metric) 61mm
Height (Metric) 104mm
Material HDPE
Mouth Narrow
No. per Pack 30
Capacity 8 oz., 250mL
Diameter (Metric) 61mm
Height (Metric) 104mm
Closure Color Natural
Closure Size 24mm
Closure Screw closure (bulk packed, unassembled)

Easily handle Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Bottles, conveniently packaged in shrink-wrap trays and ready to feed into filling lines for various reagents. High density polyethylene bottles include leakproof† polypropylene closures and rigid, lint-free trays.

Features:

  • Rigid, lint-free trays
  • Bottles can be filled in trays, providing easy handling
  • Natural high density polyethylene with polypropylene closures

Ordering Information:

Bottles are packed in shrink-wrapped trays with closures packed in separate polybags within the same carton.