Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Economy Bottles


Conduct light-duty, general purpose applications where a heavier bottle is not required with Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Bottles.

Manufacturer: thermo scientific™  NDS20850016

 View more versions of this product

 Disclaimers

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Catalog No. 03-313-1E

  • / Case of 48 Each
 

Description & Specifications

Specifications

Closure Color Amber
Neck Finish 28mm
Mouth Narrow
Closure Type Screw closure
Diameter (Metric) 73mm
Shape Round
Closure Size 28mm
Height (Metric) 170mm
Sterility Nonsterile
Certifications/Compliance Meets the requirements for light-resistant containers per USP latest edition
Description Amber Narrow-Mouth Economy Bottle
Capacity (Metric) 500mL
Closure Material Polypropylene (PP)
Material HDPE
Capacity 16 oz., 500mL
No. per Case 48
No. per Pack 48
Capacity (English) 16 oz.
Closure Liner Linerless
Diameter (Metric) Inner Neck 21mm
Color Opaque Amber

Conduct light-duty, general purpose applications where a heavier bottle is not required with Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Bottles. Lightweight versions of the original Nalgene Amber Lab Quality bottle, these high density polyethylene bottles reduce UV light transmissions to protect light-sensitive liquids.

  • Meets U.S. Pharmacopoeia 23 requirements for light transmission
  • Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles where exposure to light needs to be restricted
  • Economical and recyclable; designed for single use
  • Can be used in freezer to -100°C (-148°F) for long term storage
  • Linerless caps provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.