Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Microcentrifuge Tube Rack: ResMer™ Manufacturing Technology
Save lab space with these chemical-resistant tube racks. Stackable racks have molded letters and numbers for easy identification.
$47.34 - $47.34
Products
Catalog Number Mfr. No. Description Price Quantity    
14-809-160
thermo scientific™
N59730015
Rack Microtube Wht 1.5mL Each for $47.34
Case of 8 Each for $378.72
Description & Specifications
Save lab space with stackable Thermo Scientific™ Nalgene™ Microcentrifuge Tube Racks. These chemical-resistant racks have molded letters and numbers for easy identification. Thermo Scientific™ ResMer™ manufacturing technology retains its color and performance in harsh lab environments. Racks are useful in a wide temperature range up to 121°C and exhibit good chemical resistance.
  • Useful in a wide temperature range and exhibits good chemical resistance
  • Stack either full or empty to save benchtop space
  • Autoclavable
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.