Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Low-Profile HDPE Lowboys
Dispense most liquids, including reagents, distilled water and titrants using chemical-resistant Thermo Scientific™ Nalgene™ Low-Profile HDPE Lowboys.
$394.35 - $479.00
Specifications
Description Lowboy Low-Profile Carboy with Spigot HDPE 8 L
Material HDPE
No. per Case 4
No. per Pack 1
View More Specs
 Alert
Not recommended for use with aggressive organic solvents
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Nalgene assigns the term “leakproof”: a) To bottle/closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes; b) To bottle/closure systems of 100 or 120mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Products
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity    
02-963-4A
thermo scientific™
N23230008
8L 63mm Each for $394.35
Case of 4 Each for $1,577.40
02-963-4
thermo scientific™
23230015
15L 63mm Each for $479.00
Case of 4 Each for $1,916.00
Description & Specifications
Specifications
Description Lowboy Low-Profile Carboy with Spigot HDPE 8 L
Material HDPE
No. per Case 4
No. per Pack 1
Dispense most liquids, including reagents, distilled water and titrants using chemical-resistant Thermo Scientific™ Nalgene™ Low-Profile HDPE Lowboys. The low-profile design of these high density polyethylene lowboys fits into tight spaces on shelves and in refrigerators and can stack three high.
Features:
  • Stack up to three high to save space on benchtop or in refrigerator
  • Contour handle allows easy one-handed carrying—even when full
  • Excellent chemical resistance—can tolerate strong acids and bases for a wide variety of solutions
  • Leakproof† construction means more secure samples
Includes:
Spigot and spigot closure and four acid-proof, pressure-sensitive labels
Note:
These tests, using other liquids, may not yield the same results. To ensure safe usage, Nalgene advises customers to test bottles and closures under conditions of their planned applications. Nalgene bottles and carboys are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com